Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZO2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116498.4 | NP_192173.1 | 6 - 7 | 1300 | 65 | |
NM_178959.3 | NP_849290.1 | 6 - 7 | 1309 | 65 | ||
GenBank mRNA | AF310222.1 | - | 6 - 7 | 1256 | 65 | |
AJ010859.1 | - | 6 - 7 | 1248 | 65 | ||
AY088417.1 | - | 6 - 7 | 1231 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |