Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116594.5 | NP_567258.1 | 1 - 2 | 441 | 95 | |
GenBank mRNA | AF360372.1 | - | 1 - 2 | 117 | 95 | |
AK316725.1 | - | 1 - 2 | 158 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |