Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116718.3 | NP_567271.1 | 2 - 3 | 431 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |