Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340517.1 | NP_001329475.1 | 2 - 3 | 673 | 90 | |
NM_116727.4 | NP_567274.1 | 2 - 3 | 781 | 90 | ||
GenBank mRNA | AF424560.1 | - | 2 - 3 | 643 | 90 | |
AY062575.1 | - | 2 - 3 | 639 | 90 | ||
BT000144.1 | - | 1 - 2 | 231 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |