Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340542.1 | NP_001329202.1 | 11 - 12 | 1658 | 107 | |
NM_116758.4 | NP_192428.2 | 12 - 13 | 1955 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |