Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340542.1 | NP_001329202.1 | 2 - 3 | 380 | 126 | |
NM_116758.4 | NP_192428.2 | 3 - 4 | 677 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |