Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116791.3 | NP_567300.1 | 5 - 6 | 755 | 81 | |
GenBank mRNA | AY035106.1 | - | 5 - 6 | 660 | 81 | |
AY087807.1 | - | 5 - 6 | 677 | 81 | ||
AY113900.1 | - | 5 - 6 | 610 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |