Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340606.1 | NP_001329837.1 | 7 - 8 | 1087 | 89 | |
NM_001340607.1 | NP_001329836.1 | 8 - 9 | 1034 | 89 | ||
NM_116970.3 | NP_192640.1 | 8 - 9 | 924 | 89 | ||
GenBank mRNA | AF370534.1 | - | 8 - 9 | 854 | 89 | |
AY072503.1 | - | 8 - 9 | 800 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |