Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203413.1 | NP_001190342.1 | 2 - 3 | 324 | 107 | |
NM_121998.5 | NP_197491.2 | 2 - 3 | 242 | 107 | ||
GenBank mRNA | AY072222.1 | - | 2 - 3 | 194 | 107 | |
BT002744.1 | - | 2 - 3 | 155 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |