Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104119.3 | NP_564607.1 | 14 - 15 | 903 | 86 | |
NM_179466.3 | NP_849797.1 | 14 - 15 | 1040 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |