Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104170.3 | NP_175700.2 | 3 - 4 | 489 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |