Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMAP4K ALPHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198280.1 | NP_001185209.1 | 18 - 19 | 1966 | 124 | |
NM_001333581.1 | NP_001319212.1 | 18 - 19 | 1968 | 124 | ||
NM_104195.3 | NP_175724.2 | 18 - 19 | 1956 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |