Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3KA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036104.2 | NP_001031181.1 | 1 - 2 | 554 | 91 | |
NM_001198282.1 | NP_001185211.1 | 1 - 2 | 554 | 91 | ||
NM_001198283.1 | NP_001185212.1 | 1 - 2 | 554 | 91 | ||
NM_001333597.1 | NP_001322087.1 | 1 - 2 | 628 | 91 | ||
NM_001333598.1 | NP_001322088.1 | 1 - 2 | 579 | 91 | ||
NM_104235.4 | NP_564635.1 | 1 - 2 | 583 | 91 | ||
NM_179472.3 | NP_849803.1 | 1 - 2 | 628 | 91 | ||
GenBank mRNA | AY140005.1 | - | 1 - 2 | 479 | 91 | |
BT002598.1 | - | 1 - 2 | 354 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |