Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRX-LIKE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160950.2 | NP_001154422.1 | 4 - 5 | 1038 | 77 | |
NM_104296.5 | NP_175820.2 | 5 - 6 | 993 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |