Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104315.5 | NP_175839.2 | 12 - 13 | 918 | 76 | |
GenBank mRNA | AK226435.1 | - | 12 - 13 | 818 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |