Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124016.2 | NP_001117488.1 | 2 - 3 | 575 | 143 | |
NM_001160953.1 | NP_001154425.1 | 3 - 4 | 353 | 143 | ||
NM_001333645.1 | NP_001320514.1 | 2 - 3 | 568 | 143 | ||
NM_001333646.1 | NP_001320513.1 | 1 - 2 | 261 | 143 | ||
NM_104317.3 | NP_564658.2 | 3 - 4 | 398 | 143 | ||
NM_202296.2 | NP_974025.1 | 3 - 4 | 398 | 143 | ||
NM_202297.3 | NP_974026.1 | 3 - 4 | 484 | 143 | ||
NM_202298.3 | NP_974027.1 | 3 - 4 | 484 | 143 | ||
GenBank mRNA | AK316931.1 | - | 3 - 4 | 391 | 143 | |
AY085569.1 | - | 3 - 4 | 353 | 143 | ||
AY099578.1 | - | 3 - 4 | 346 | 143 | ||
BT002159.1 | - | 3 - 4 | 317 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |