Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198295.1 | NP_001185224.1 | 11 - 12 | 735 | 118 | |
NM_001198296.1 | NP_001185225.1 | 11 - 12 | 670 | 118 | ||
NM_001333653.1 | NP_001319226.1 | 10 - 11 | 1078 | 118 | ||
NM_104329.5 | NP_175853.1 | 11 - 12 | 831 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |