Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333687.1 | NP_001320298.1 | 3 - 4 | 851 | 112 | |
NM_104382.5 | NP_175905.2 | 4 - 5 | 754 | 112 | ||
GenBank mRNA | AK227122.1 | - | 4 - 5 | 755 | 112 | |
AY093241.1 | - | 4 - 5 | 711 | 112 | ||
BT006281.1 | - | 4 - 5 | 674 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |