Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198317.1 | NP_001185246.1 | 5 - 6 | 732 | 107 | |
NM_104467.5 | NP_175986.2 | 5 - 6 | 719 | 107 | ||
GenBank mRNA | AY072140.1 | - | 5 - 6 | 716 | 107 | |
AY122975.1 | - | 5 - 6 | 622 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |