Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104512.3 | NP_176029.1 | 1 - 2 | 199 | 75 | |
GenBank mRNA | AY072220.1 | - | 1 - 2 | 188 | 75 | |
AY085815.1 | - | 1 - 2 | 194 | 75 | ||
AY096599.1 | - | 1 - 2 | 123 | 75 | ||
M95795.1 | - | 1 - 2 | 182 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |