Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104541.4 | NP_176057.1 | 2 - 3 | 459 | 77 | |
GenBank mRNA | AK221639.1 | - | 2 - 3 | 304 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |