Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104624.4 | NP_564734.1 | 7 - 8 | 1607 | 94 | |
GenBank mRNA | AB008021.1 | - | 7 - 8 | 1501 | 94 | |
AY099643.1 | - | 7 - 8 | 1517 | 94 | ||
BT000239.1 | - | 7 - 8 | 1473 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |