Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CW7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333852.1 | NP_001319271.1 | 2 - 3 | 532 | 81 | |
NM_001333853.1 | NP_001321061.1 | 2 - 3 | 532 | 81 | ||
NM_001333854.1 | NP_001321060.1 | 2 - 3 | 532 | 81 | ||
NM_104645.4 | NP_564743.1 | 2 - 3 | 532 | 81 | ||
NM_179496.2 | NP_849827.1 | 2 - 3 | 517 | 81 | ||
NM_179497.1 | NP_849828.1 | 2 - 3 | 430 | 81 | ||
GenBank mRNA | AB028226.1 | - | 2 - 3 | 337 | 81 | |
AK227804.1 | - | 2 - 3 | 414 | 81 | ||
BT004825.1 | - | 2 - 3 | 225 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |