Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CW14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104658.4 | NP_564750.1 | 4 - 5 | 1111 | 137 | |
GenBank mRNA | AB028233.1 | - | 4 - 5 | 1111 | 137 | |
BT004250.1 | - | 4 - 5 | 837 | 137 | ||
BT005503.1 | - | 4 - 5 | 757 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |