Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CW14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104658.4 | NP_564750.1 | 8 - 9 | 1718 | 110 | |
GenBank mRNA | AB028233.1 | - | 8 - 9 | 1718 | 110 | |
BT004250.1 | - | 8 - 9 | 1444 | 110 | ||
BT005503.1 | - | 8 - 9 | 1364 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |