Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333894.1 | NP_001320517.1 | 6 - 7 | 974 | 120 | |
NM_001333895.1 | NP_001320518.1 | 6 - 7 | 738 | 120 | ||
NM_104731.5 | NP_176247.2 | 7 - 8 | 870 | 120 | ||
GenBank mRNA | AK229327.1 | - | 7 - 8 | 853 | 120 | |
BT011005.1 | - | 7 - 8 | 720 | 120 | ||
BT015384.1 | - | 7 - 8 | 682 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |