Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198351.1 | NP_001185280.1 | 3 - 4 | 336 | 70 | |
NM_104833.3 | NP_176347.1 | 2 - 3 | 464 | 70 | ||
GenBank mRNA | AY059879.1 | - | 2 - 3 | 293 | 70 | |
AY093370.1 | - | 2 - 3 | 205 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |