Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BETA-VPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104948.4 | NP_176458.1 | 7 - 8 | 1198 | 64 | |
GenBank mRNA | AF367254.1 | - | 7 - 8 | 1102 | 64 | |
AY059156.1 | - | 7 - 8 | 1072 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |