Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BETA-VPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104948.4 | NP_176458.1 | 8 - 9 | 1399 | 71 | |
GenBank mRNA | AF367254.1 | - | 8 - 9 | 1303 | 71 | |
AY059156.1 | - | 8 - 9 | 1273 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |