Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104974.4 | NP_564804.1 | 1 - 2 | 735 | 130 | |
GenBank mRNA | AF348575.1 | - | 1 - 2 | 537 | 130 | |
AY054691.1 | - | 1 - 2 | 575 | 130 | ||
BT008906.1 | - | 1 - 2 | 537 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |