Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104990.4 | NP_176500.1 | 3 - 4 | 1022 | 100 | |
GenBank mRNA | AB195671.1 | - | 3 - 4 | 608 | 100 | |
BT004076.1 | - | 3 - 4 | 698 | 100 | ||
BT005121.1 | - | 3 - 4 | 608 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |