Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202746.1 | NP_001189675.1 | 17 - 18 | 2175 | 124 | |
NM_001336522.1 | NP_001324371.1 | 11 - 12 | 1148 | 124 | ||
NM_129020.3 | NP_181013.1 | 16 - 17 | 2440 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |