Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202746.1 | NP_001189675.1 | 6 - 7 | 1122 | 74 | |
NM_129020.3 | NP_181013.1 | 5 - 6 | 1387 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |