Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084537.1 | NP_001078006.1 | 5 - 6 | 614 | 72 | |
NM_129053.6 | NP_181046.1 | 5 - 6 | 651 | 72 | ||
GenBank mRNA | BT025773.1 | - | 5 - 6 | 487 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |