Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036408.3 | NP_001031485.1 | 11 - 12 | 1458 | 117 | |
NM_129064.3 | NP_181056.2 | 10 - 11 | 1424 | 117 | ||
GenBank mRNA | AY496700.1 | - | 11 - 12 | 1417 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |