Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105114.3 | NP_176621.1 | 6 - 7 | 1807 | 70 | |
GenBank mRNA | AF372940.1 | - | 6 - 7 | 1636 | 70 | |
AY143945.1 | - | 6 - 7 | 1502 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |