Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105130.4 | NP_176636.1 | 4 - 5 | 916 | 110 | |
GenBank mRNA | BT002015.1 | - | 4 - 5 | 858 | 110 | |
BT010544.1 | - | 4 - 5 | 816 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |