Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105138.5 | NP_176644.2 | 1 - 2 | 486 | 103 | |
GenBank mRNA | AB493519.1 | - | 1 - 2 | 56 | 103 | |
AY081307.1 | - | 1 - 2 | 477 | 103 | ||
BT010383.1 | - | 1 - 2 | 56 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |