Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105138.5 | NP_176644.2 | 2 - 3 | 519 | 99 | |
GenBank mRNA | AB493519.1 | - | 2 - 3 | 89 | 99 | |
AY081307.1 | - | 2 - 3 | 510 | 99 | ||
BT010383.1 | - | 2 - 3 | 89 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |