Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334199.1 | NP_001322349.1 | 7 - 8 | 1047 | 135 | |
NM_105199.3 | NP_564846.1 | 7 - 8 | 892 | 135 | ||
GenBank mRNA | AY045845.1 | - | 7 - 8 | 660 | 135 | |
AY117153.1 | - | 7 - 8 | 651 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |