Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_110977.3 | NP_186761.1 | 3 - 4 | 1021 | 74 | |
GenBank mRNA | AY094438.1 | - | 3 - 4 | 759 | 74 | |
BT002753.1 | - | 3 - 4 | 727 | 74 | ||
U83500.1 | - | 3 - 4 | 747 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |