Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340791.1 | NP_001319913.1 | 6 - 7 | 1089 | 81 | |
NM_001340792.1 | NP_001319914.1 | 6 - 7 | 1072 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |