Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117355.1 | NP_193022.1 | 1 | 262 | 76 | |
GenBank mRNA | DQ056648.1 | - | 1 | 262 | 76 |
Gene Symbol: AT4G12850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117354.3 | NP_193021.2 | 2 | 1011 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |