Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAKRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117492.2 | NP_567423.1 | 7 - 8 | 1038 | 60 | |
GenBank mRNA | AF193767.1 | - | 7 - 8 | 832 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |