Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117499.5 | NP_193158.2 | 1 - 2 | 317 | 82 | |
GenBank mRNA | DQ059122.1 | - | 1 - 2 | 186 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |