Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340945.1 | NP_001329483.1 | 3 - 4 | 930 | 74 | |
NM_117535.5 | NP_193191.2 | 3 - 4 | 930 | 74 | ||
GenBank mRNA | AB493680.1 | - | 3 - 4 | 590 | 74 | |
AF334718.1 | - | 3 - 4 | 590 | 74 | ||
AK228403.1 | - | 3 - 4 | 686 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |