Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OASA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203803.1 | NP_001190732.1 | 10 - 11 | 1165 | 72 | |
NM_001203804.1 | NP_001190733.1 | 10 - 11 | 1168 | 72 | ||
NM_001340975.1 | NP_001328013.1 | 10 - 11 | 1347 | 72 | ||
NM_117574.4 | NP_193224.1 | 10 - 11 | 1152 | 72 | ||
NM_179055.3 | NP_849386.1 | 10 - 11 | 1022 | 72 | ||
GenBank mRNA | BT025878.1 | - | 9 - 10 | 874 | 72 | |
X80376.2 | - | 935 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |