Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036570.1 | NP_001031647.1 | 14 - 15 | 2228 | 81 | |
NM_001084926.1 | NP_001078395.1 | 13 - 14 | 2165 | 81 | ||
NM_001084927.2 | NP_001078396.1 | 12 - 13 | 2217 | 81 | ||
NM_001125515.1 | NP_001118987.1 | 12 - 13 | 2110 | 81 | ||
NM_001341051.1 | NP_001328869.1 | 12 - 13 | 2336 | 81 | ||
NM_117643.4 | NP_193288.2 | 11 - 12 | 1966 | 81 | ||
NM_179060.4 | NP_849391.2 | 13 - 14 | 2204 | 81 | ||
GenBank mRNA | AK317187.1 | - | 13 - 14 | 2193 | 81 | |
AK317578.1 | - | 11 - 12 | 1884 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |