Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341063.1 | NP_001329006.1 | 7 - 8 | 1233 | 146 | |
NM_001341064.1 | NP_001329005.1 | 7 - 8 | 989 | 146 | ||
NM_001341065.1 | NP_001329007.1 | 5 - 6 | 704 | 146 | ||
NM_117648.8 | NP_193292.3 | 8 - 9 | 1026 | 146 | ||
GenBank mRNA | EU915246.1 | - | 8 - 9 | 1026 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |