Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341097.1 | NP_001319958.1 | 3 - 4 | 615 | 138 | |
NM_117729.8 | NP_193367.7 | 2 - 3 | 567 | 138 | ||
GenBank mRNA | AF465831.1 | - | 2 - 3 | 500 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |